1-(3-Hydroxypyridin-4-yl)ethanone 97%

Supplier: Ambeed
Warning

A134050-100MG A134050-250MG
103801-962EA 136 USD
103801-962 77883-081 77883-082
1-(3-Hydroxypyridin-4-yl)ethanone 97%
1-(3-Hydroxypyridin-4-yl)ethanone

Precaución: Inert atmosphere, room temperature

Formula: C₇H₇NO₂
Melting Pt: 52…54 °C
Flash Pt: 157.291 °C
Storage Temperature: Ambient
MDL Number: MFCD18256467
CAS Number: 30152-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR