1-(3-Hydroxypyridin-2-yl)ethanone 98%

Supplier: Ambeed
Warning

A102278-100MG A102278-10G A102278-25G A102278-1G A102278-5G
103796-350EA 96 USD
103796-350 103822-042 76970-208 76970-210 76970-212 77308-920 77308-922
1-(3-Hydroxypyridin-2-yl)ethanone 98%
1-(3-Hydroxypyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.136 g/mol
Boiling Pt: 311.5 °C (760 mm Hg)
Flash Pt: 142.217 °C
Storage Temperature: Ambient
MDL Number: MFCD08062669
CAS Number: 13210-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR