1-(3-Chloropyridin-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A101065-250MG A101065-1G
103795-788EA 43 USD
103795-788 103821-478 76982-170 77296-974
1-(3-Chloropyridin-2-yl)ethanone 95%
1-(3-Chloropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656222
CAS Number: 131109-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR