1-(3-Chloro-2-hydroxyphenyl)ethanone 98%

Supplier: Ambeed
Warning

A228092-1G A228092-5G A228092-10G
103819-358EA 45 USD
103819-358 103850-266 77089-058 77089-060 77877-735
1-(3-Chloro-2-hydroxyphenyl)ethanone 98%
1-(3-Chloro-2-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
Boiling Pt: 247.1 °C (760 mm Hg)
Melting Pt: 80 °C
Flash Pt: 103.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00100492
CAS Number: 3226-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR