1-(3-Bromopropyl)pyrrolidine hydrobromide 97%

Supplier: Ambeed
Warning

A169085-1G A169085-100MG A169085-250MG
103806-350EA 176 USD
103806-350 77081-678 77110-922 77110-924
1-(3-Bromopropyl)pyrrolidine hydrobromide 97%
1-(3-Bromopropyl)pyrrolidine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₁₅Br₂N
MW: 273 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07787235
CAS Number: 88806-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR