(3-Bromobenzyl)methylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Bromophenyl)-N-methylmethanamine, 3-Bromo-N-methylbenzylamine

A172869-1G A172869-5G A172869-100G A172869-25G
103806-760EA 13 USD
103806-760 103845-052 77082-266 77082-268 77111-088 77301-232
(3-Bromobenzyl)methylamine 98%
(3-Bromobenzyl)methylamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 231…232 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04507522
CAS Number: 67344-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR