1-(3-Aminopropyl)azepan-2-one 95%

Supplier: Ambeed
Warning

A849114-250MG A849114-1G
103841-228EA 624 USD
103841-228 76728-836 77308-450
1-(3-Aminopropyl)azepan-2-one 95%
1-(3-Aminopropyl)azepan-2-one

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O
MW: 170.25 g/mol
Boiling Pt: 326 °C (760 mm Hg)
Flash Pt: 150.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036060
CAS Number: 24566-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR