3'-Amino-2'-hydroxyacetophenone 98%

Supplier: Ambeed
Warning

A134415-10G A134415-100G A134415-500G
76811-402EA 7 USD
76811-402 77670-930 77883-083
3'-Amino-2'-hydroxyacetophenone 98%
3'-Amino-2'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₉NO₂
Boiling Pt: 287.2 °C (760 mm Hg)
Flash Pt: 127.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03428566
CAS Number: 70977-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR