3',5'-Dimethylacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Dimethylphenyl)ethanone, 1-(3,5-Dimethylphenyl)ethan-1-one

A113444-100G A113444-10G A113444-25G A113444-1G A113444-5G
103798-234EA 5 USD
103798-234 103824-076 76687-690 76687-692 76687-694 77310-890 77310-892
3',5'-Dimethylacetophenone 98%
3',5'-Dimethylacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 98…100 °C (760 mm Hg)
Melting Pt: 20 °C
Flash Pt: 93.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01075693
CAS Number: 5379-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR