1-(3,5-Dichlorophenyl)propan-1-one 95%

Supplier: Ambeed
Warning

A113897-100MG A113897-250MG A113897-1G
103798-312EA 87 USD
103798-312 103824-154 76982-862 76982-864 77310-968
1-(3,5-Dichlorophenyl)propan-1-one 95%
1-(3,5-Dichlorophenyl)propan-1-one

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617650
CAS Number: 92821-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR