3',5'-Dichloroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Dichlorophenyl)ethanone

A526896-100G A526896-5G A526896-1G A526896-25G
103828-540EA 6 USD
103828-540 103839-652 76774-700 76843-662 77307-724 77307-726
3',5'-Dichloroacetophenone 97%
3',5'-Dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 285.4 °C (760 mm Hg)
Melting Pt: 26…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00045189
CAS Number: 14401-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR