3',5'-Dichloro-2,2,2-trifluoroacetophenone 95%
Supplier: Ambeed
|
Warning
|
A225784-100G
A225784-100MG
A225784-10G
A225784-250MG
A225784-1G
A225784-25G
A225784-5G
103819-198EA
8
USD
103819-198
103850-098
76874-104
76874-106
76874-108
76874-110
77088-804
77088-806
77088-808
3',5'-Dichloro-2,2,2-trifluoroacetophenone 95%
3',5'-Dichloro-2,2,2-trifluoroacetophenone
Precaución: for Research Use Only
Formula:
C₈H₃Cl₂F₃O Boiling Pt: 265 °C (760 mm Hg) Flash Pt: 114 °C Storage Temperature: Ambient |
MDL Number:
MFCD01319996 CAS Number: 130336-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...