3',5'-Dibromoacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Dibromophenyl)ethanone

A249272-100G A249272-5G A249272-25G
103811-862EA 8 USD
103811-862 103851-700 77031-474 77091-046 77304-476
3',5'-Dibromoacetophenone 97%
3',5'-Dibromoacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Br₂O
MW: 277.9400 g/mol
Boiling Pt: 318 °C (760 mm Hg)
Flash Pt: 118.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00078665
CAS Number: 14401-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR