3',4'-Dichloroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-dichlorophenyl)ethanone

A307007-100G A307007-10G A307007-500G A307007-25G
103835-668EA 5 USD
103835-668 76772-734 76772-736 76772-738 76838-402
3',4'-Dichloroacetophenone 98%
3',4'-Dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 264.8 °C (760 mm Hg)
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00000553
CAS Number: 2642-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR