1-(3-(Trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butane-1,3-dione 98%

Supplier: Ambeed
Warning

Synonyms: rac-Sitagliptin 3-Ketone

A107286-100G A107286-10G A107286-1G A107286-25G A107286-5G
103822-928EA 16 USD
103822-928 77283-562 77283-564 77283-566 77283-568 77309-778
1-(3-(Trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butane-1,3-dione 98%
1-(3-(Trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butane-1,3-dione

Precaución: for Research Use Only

Formula: C₁₆H₁₂F₆N₄O₂
MW: 406.28 g/mol
Boiling Pt: 513.902 °C (760 mm Hg)
Melting Pt: 92.5 °C
Flash Pt: 264.598 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11111443
CAS Number: 764667-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR