1-Pyrimidin-2-ylethanone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Pyrimidinyl)ethanone, 2-Acetylpyrimidine

A165888-10G A165888-250MG A165888-5G A165888-1G A165888-25G
103806-008EA 15 USD
103806-008 103844-262 76799-192 76799-194 77300-802 77320-484 77320-486
1-Pyrimidin-2-ylethanone 97%
1-Pyrimidin-2-ylethanone

Precaución: for Research Use Only

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 254.4 °C (760 mm Hg)
Melting Pt: 52 °C
Flash Pt: 111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08361636
CAS Number: 53342-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR