1-(2-Phenylpropan-2-yl)piperidine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-2-(1-piperidinyl)propane

A127889-100MG A127889-1G A127889-250MG A127889-5G
103801-034EA 142 USD
103801-034 77009-772 77313-306 77683-135 77889-884
1-(2-Phenylpropan-2-yl)piperidine 98%, Ambeed.Inc
1-(2-Phenylpropan-2-yl)piperidine

Precaución: For research use only.

Formula: C₁₄H₂₁N
MW: 203.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00870884
CAS Number: 92321-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR