2'-Nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone

A138850-100G A138850-10G A138850-500G A138850-5G A138850-25G A138850-1000G
76972-496EA 50.93 USD
76972-496 76972-498 76972-500 77299-026 77315-730 77670-990
2'-Nitroacetophenone 98%
2'-Nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 286.4 °C (760 mm Hg)
Melting Pt: 23…27 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007144
CAS Number: 577-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR