1-(2-N-Boc-aminoethyl)piperazine 97%

Supplier: Ambeed
Warning

A327041-100MG A327041-250MG
103825-454EA 45 USD
103825-454 103836-406 76773-016 77598-426
1-(2-N-Boc-aminoethyl)piperazine 97%
1-(2-N-Boc-aminoethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₃N₃O₂
Boiling Pt: 357.2 °C (760 mm Hg)
Flash Pt: 169.8 °C
Storage Temperature: Freezer
MDL Number: MFCD02683050
CAS Number: 140447-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR