2-Methoxy-N-methylbenzylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Methoxyphenyl)-N-methylmethanamine

A115874-250MG A115874-1G
103798-642EA 25 USD
103798-642 103824-504 77257-934 77311-328
2-Methoxy-N-methylbenzylamine 95%
2-Methoxy-N-methylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.209 g/mol
Boiling Pt: 234…235 °C (760 mm Hg)
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025755
CAS Number: 6851-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR