1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-[1-(2-Methoxyethyl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid Pinacol ester, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester

A213247-100MG A213247-250MG A213247-1G
103818-280EA 28 USD
103818-280 103848-986 76975-590 76975-592 77087-490
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₂H₂₁BN₂O₃
Melting Pt: 53…57 °C
Flash Pt: 169.4 °C
Storage Temperature: Freezer
MDL Number: MFCD12405649
CAS Number: 847818-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR