4-Methylpiperazine-1-ethanol 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxyethyl)-4-methylpiperazine

A444003-1G A444003-25G A444003-5G A444003-100G A444003-10G A444003-500G
103827-704EA 5 USD
103827-704 103838-772 76842-550 76842-552 76842-554 77116-150 77116-152 77116-154
4-Methylpiperazine-1-ethanol 97%
4-Methylpiperazine-1-ethanol

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
Boiling Pt: 234.9 °C (760 mm Hg)
Melting Pt: 39…41 °C
Flash Pt: 106.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03840691
CAS Number: 5464-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR