1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Hydroxyethyl)-3-methyl-1H-imidazol-3-ium tetrafluoroborate
A103039-100G
A103039-10G
A103039-25G
A103039-1G
A103039-5G
103796-472EA
6
USD
103796-472
103822-174
77093-598
77093-600
77093-602
77283-028
77283-030
1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate 96%
1-(2-Hydroxyethyl)-3-methylimidazolium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₆H₁₁BF₄N₂O MW: 213.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12761438 CAS Number: 374564-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...