1-(2-Hydroxy-6-methylphenyl)ethanone 95%

Supplier: Ambeed
Warning

A116491-100MG A116491-250MG A116491-1G A116491-5G
103798-740EA 65 USD
103798-740 103824-606 76971-224 76971-226 77297-470 77297-472
1-(2-Hydroxy-6-methylphenyl)ethanone 95%
1-(2-Hydroxy-6-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18397234
CAS Number: 41085-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR