2'-Hydroxy-6'-methoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxy-6-methoxyphenyl)ethanone

A321158-10G A321158-250MG A321158-1G A321158-25G A321158-5G
103825-266EA 6 USD
103825-266 103836-200 76693-790 76693-792 76839-094 76839-096 76839-098
2'-Hydroxy-6'-methoxyacetophenone 98%
2'-Hydroxy-6'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18
Boiling Pt: 259.5 °C (760 mm Hg)
Melting Pt: 56…59 °C
Flash Pt: 108.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008732
CAS Number: 703-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR