2'-Hydroxy-5'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxy-5-nitrophenyl)ethanone

A216114-1G A216114-5G A216114-250MG
103818-538EA 6 USD
103818-538 103849-250 77087-824 77087-826 77237-558
2'-Hydroxy-5'-nitroacetophenone 98%
2'-Hydroxy-5'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.1500 g/mol
Boiling Pt: 285.5 °C (760 mm Hg)
Melting Pt: 100…104 °C
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00463816
CAS Number: 1450-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR