1-(2-Hydroxy-5-methylphenyl)-3-phenylpropane-1,3-dione 97%

Supplier: Ambeed
Warning

A172652-100MG A172652-250MG A172652-1G A172652-5G
103806-734EA 236 USD
103806-734 103845-026 77191-468 77191-470 77301-208 77301-210
1-(2-Hydroxy-5-methylphenyl)-3-phenylpropane-1,3-dione 97%
1-(2-Hydroxy-5-methylphenyl)-3-phenylpropane-1,3-dione

Precaución: Sealed in dry, at room temperature.

Formula: C₁₆H₁₄O₃
MW: 254.28 g/mol
Boiling Pt: 455.7 °C (760 mm Hg)
Melting Pt: 92…94 °C
Flash Pt: 243.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00209591
CAS Number: 29976-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR