2'-Hydroxy-4'-methoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol

A173012-10G A173012-1G A173012-500G A173012-5G A173012-25G A173012-100G A173012-1000G
103845-070EA 6 USD
103845-070 76974-098 76974-100 76974-102 77082-296 77301-238 77593-084 77671-456
2'-Hydroxy-4'-methoxyacetophenone 98%
2'-Hydroxy-4'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 154
Melting Pt: 51
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008730
CAS Number: 552-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR