(2-Fluorophenyl)acetone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluorophenylacetone, 1-(2-Fluorophenyl)propan-2-one, 1-(2-Fluorophenyl)-2-propanone

A620941-5G A620941-100G A620941-10G A620941-1G A620941-25G
103840-216EA 52 USD
103840-216 76844-386 77266-856 77266-858 77307-970 77307-972
(2-Fluorophenyl)acetone 98%
(2-Fluorophenyl)acetone

Precaución: for Research Use Only

Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 266.2 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000325
CAS Number: 2836-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR