2-Fluoro-N-methylbenzylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine

A282408-25G A282408-10G A282408-1G A282408-5G
103813-590EA 7 USD
103813-590 103853-542 76837-238 77141-194 77305-432 77305-434
2-Fluoro-N-methylbenzylamine 98%
2-Fluoro-N-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD04629633
CAS Number: 399-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR