2'-Fluoroacetophenone 98%

Supplier: Ambeed
Warning

A257107-100G A257107-10G A257107-500G A257107-1000G A257107-25G A257107-5G
103809-824EA 171 USD
103809-824 103852-156 76815-886 76815-888 76815-890 77091-660 77091-662 77304-692
2'-Fluoroacetophenone 98%
2'-Fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇FO
Boiling Pt: 180.8 °C (760 mm Hg)
Melting Pt: 26…27 °C
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000320
CAS Number: 445-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR