2'-Fluoro-6'-hydroxyacetophenone 98%
Supplier: Ambeed
|
Warning
|
A144864-100G
A144864-100MG
A144864-10G
A144864-250MG
A144864-1G
A144864-25G
A144864-5G
103803-462EA
24
USD
103803-462
103841-540
77109-986
77109-988
77109-990
77109-992
77316-794
77316-796
77316-798
2'-Fluoro-6'-hydroxyacetophenone 98%
2'-Fluoro-6'-hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇FO₂ Boiling Pt: 224 °C (760 mm Hg) Flash Pt: 89.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01090995 CAS Number: 93339-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...