2'-Fluoro-5'-nitroacetophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Fluoro-5-nitrophenyl)ethanone
A490255-1G
A490255-100G
A490255-25G
A490255-10G
103828-212EA
5
USD
103828-212
103839-284
76843-246
77196-880
77196-882
77747-008
2'-Fluoro-5'-nitroacetophenone 98%
2'-Fluoro-5'-nitroacetophenone
Precaución: for Research Use Only
Formula:
C₈H₆FNO₃ MW: 183.1400 g/mol Boiling Pt: 241 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00277867 CAS Number: 79110-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...