1-(2-Chloropyrimidin-5-yl)ethanone 95%

Supplier: Ambeed
Warning

A141686-10G A141686-250MG
103803-038EA 45 USD
103803-038 76871-246 76871-248
1-(2-Chloropyrimidin-5-yl)ethanone 95%
1-(2-Chloropyrimidin-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Boiling Pt: 307.9±15.0 °C (760 mm Hg)
Melting Pt: 92 °C
Flash Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697144
CAS Number: 110100-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR