1-(2-Chloropyrimidin-4-yl)ethanone 97%

Supplier: Ambeed
Warning

A278276-100MG A278276-250MG A278276-1G
103813-408EA 146 USD
103813-408 103853-350 76827-472 76827-474 77093-262
1-(2-Chloropyrimidin-4-yl)ethanone 97%
1-(2-Chloropyrimidin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD22543973
CAS Number: 1312535-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR