2'-Chloropropiophenone 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chlorophenyl Ethyl Ketone, 1-(2-Chlorophenyl)propan-1-one

A402021-10G A402021-250MG A402021-1G A402021-25G A402021-5G
103827-070EA 26 USD
103827-070 103838-106 76876-908 76876-910 77307-078 77307-080 77307-082
2'-Chloropropiophenone 95%
2'-Chloropropiophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787256
CAS Number: 6323-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR