1-(2-Chloroethyl)pyrrolidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-Chloroethyl)pyrrolidine hydrochloride

A138546-100G A138546-10G A138546-500G A138546-25G A138546-5G
76811-654EA 57 USD
76811-654 76811-656 76811-658 77298-998 77299-000
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClN
MW: 170.08 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00012718
CAS Number: 7250-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR