1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Chloroethyl)hexamethyleneimine Hydrochloride, 1-(2-Chloroethyl)azepane hydrochloride
A529422-25G
A529422-100G
A529422-10G
A529422-1G
103828-558EA
14
USD
103828-558
103839-668
76843-684
77197-320
77197-322
77307-732
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride 98%
1-(2-Chloroethyl)-1H-hexahydroazepine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₆ClN MW: 198.13 g/mol Boiling Pt: 213.3 °C (760 mm Hg) Melting Pt: 208…210 °C Flash Pt: 82.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012842 CAS Number: 26487-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...