1-(2-Chloroethyl)-4-methylpiperazine hydrochloride 95%

Supplier: Ambeed
Warning

A137151-10G A137151-250MG A137151-1G A137151-5G
103802-422EA 39 USD
103802-422 76871-004 76871-006 77298-896 77315-376
1-(2-Chloroethyl)-4-methylpiperazine hydrochloride 95%
1-(2-Chloroethyl)-4-methylpiperazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂·ClH
MW: 199.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031872
CAS Number: 126055-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR