1-(2-Chloroethyl)imidazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Chloroethyl)-1H-imidazole hydrochloride

A160979-10G A160979-250MG A160979-1G A160979-5G
103805-438EA 25 USD
103805-438 103843-676 76720-126 76720-128 77300-506 77300-508
1-(2-Chloroethyl)imidazole hydrochloride 95%
1-(2-Chloroethyl)imidazole hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈Cl₂N₂
MW: 167.04 g/mol
Melting Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00799332
CAS Number: 18994-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR