1-(2-Chlorobenzoyl)piperidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A210303-1G A210303-5G
103817-994EA 30 USD
103817-994 103848-688 77087-056 77087-058
1-(2-Chlorobenzoyl)piperidine-4-carboxylic acid 98%
1-(2-Chlorobenzoyl)piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₄ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD02070180
CAS Number: 352673-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR