4-Nitrophenethyl bromide 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene

A104989-100G A104989-25G A104989-500G A104989-5G
103807-922EA 323 USD
103807-922 103822-536 76687-202 76687-204 76687-206 76687-208 77283-316
4-Nitrophenethyl bromide 98%
4-Nitrophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Boiling Pt: 309.6 °C (760 mm Hg)
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00007386
CAS Number: 5339-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR