1-(2-Bromoethyl)-4-methylpiperazine dihydrobromide 97%

Supplier: Ambeed
Warning

A115956-100MG A115956-250MG A115956-1G A115956-5G
103798-650EA 37 USD
103798-650 103824-514 76995-522 76995-524 77297-408 77297-410
1-(2-Bromoethyl)-4-methylpiperazine dihydrobromide 97%
1-(2-Bromoethyl)-4-methylpiperazine dihydrobromide

Precaución: for Research Use Only

Formula: C₇H₁₇Br₃N₂
MW: 368.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12068401
CAS Number: 5845-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR