4-Chlorophenethyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(2-Bromoethyl)-4-chlorobenzene

A321078-100G A321078-25G A321078-5G
103825-262EA 9 USD
103825-262 103836-194 76693-788 76839-082 76839-084
4-Chlorophenethyl bromide 97%
4-Chlorophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.51 g/mol
Boiling Pt: 239.3 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00634271
CAS Number: 6529-53-9
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR