4-Trifluoromethylphenethyl bromide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Bromoethyl)-4-(trifluoromethyl)benzene
A260192-250MG
A260192-1G
A260192-5G
103812-480EA
52
USD
103812-480
103852-362
76988-170
77304-778
77304-780
4-Trifluoromethylphenethyl bromide 95%
4-Trifluoromethylphenethyl bromide
Precaución: for Research Use Only
Formula:
C₉H₈BrF₃ Boiling Pt: 218.0±35.0 °C (760 mm Hg) Flash Pt: 99.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD10567022 CAS Number: 130365-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...