6'-Bromo-2'-hydroxyacetophenone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Bromo-6-hydroxyphenyl)ethanone, 2-Acetyl-3-bromophenol, 1-(2-Bromo-6-hydroxyphenyl)ethan-1-one
A235940-250MG
A235940-1G
A235940-25G
103811-050EA
87
USD
103811-050
103850-820
76815-280
77089-846
77089-848
6'-Bromo-2'-hydroxyacetophenone 97%
6'-Bromo-2'-hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇BrO₂ MW: 215.044 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD13193630 CAS Number: 55736-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...