1-(2-Bromo-4-hydroxyphenyl)ethanone 98%

Supplier: Ambeed
Warning

A112537-100MG A112537-250MG
103798-032EA 33 USD
103798-032 103823-868 77257-740 77257-742
1-(2-Bromo-4-hydroxyphenyl)ethanone 98%
1-(2-Bromo-4-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.044 g/mol
Boiling Pt: 344.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03428552
CAS Number: 61791-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR