1-(2-Amino-3-pyridinyl)-1-ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Aminopyridin-3-yl)ethanone, 3-Acetyl-2-aminopyridine

A169735-1G A169735-5G A169735-100G A169735-100MG A169735-10G A169735-250MG
103806-402EA 18 USD
103806-402 103844-666 77081-762 77081-764 77163-486 77163-488 77163-490 77163-492
1-(2-Amino-3-pyridinyl)-1-ethanone 97%
1-(2-Amino-3-pyridinyl)-1-ethanone

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 283.8 °C (760 mm Hg)
Melting Pt: 139…140 °C
Flash Pt: 125.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02258895
CAS Number: 65326-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR