1-(2-Aminoethyl)-2-imidazolidone 97%

Supplier: Ambeed
Danger

Synonyms: 1-(2-Aminoethyl)imidazolidin-2-one

A139435-10G A139435-250MG A139435-1G A139435-5G
103802-742EA 29 USD
103802-742 76984-126 76984-128 77315-836 77315-838
1-(2-Aminoethyl)-2-imidazolidone 97%
1-(2-Aminoethyl)-2-imidazolidone

Precaución: for Research Use Only

Formula: C₅H₁₁N₃O
MW: 129.16 g/mol
Boiling Pt: 363.1 °C (760 mm Hg)
Flash Pt: 173.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00086348
CAS Number: 6281-42-1
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR