1-(2-Amino-5-fluorophenyl)ethanone 97%

Supplier: Ambeed
Warning

A125240-100MG A125240-10G A125240-250MG A125240-25G A125240-1G A125240-5G
103800-650EA 10 USD
103800-650 76870-454 76870-456 76870-458 76870-460 77312-798 77312-800
1-(2-Amino-5-fluorophenyl)ethanone 97%
1-(2-Amino-5-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09835333
CAS Number: 2343-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR